Anti-CCL15 Goat Polyclonal Antibody

Supplier: R&D Systems
AF628
220012-458EA 649.32 USD
220012-458
Anti-CCL15 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CCL15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR