Anti-CMV UL146/vCXC1 Goat Polyclonal Antibody

Supplier: R&D Systems
AF620
220012-438EA 634.86 USD
220012-438
Anti-CMV UL146/vCXC1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CMV UL146/vCXC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR