Anti-PF4 Goat Polyclonal Antibody

Supplier: R&D Systems
AF595
220012-351EA 663.78 USD
220012-351
Anti-PF4 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR