Anti-IL21 Goat Polyclonal Antibody

Supplier: R&D Systems
AF594
220012-343EA 721.63 USD
220012-343
Anti-IL21 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR