Anti-PDZK1 Goat Polyclonal Antibody

Supplier: R&D Systems
AF5780
220012-269EA 620.4 USD
220012-269
Anti-PDZK1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PDZK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR