Anti-APBA3 Sheep Polyclonal Antibody

Supplier: R&D Systems
AF5689
220012-221EA 649.32 USD
220012-221
Anti-APBA3 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: APBA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR