Anti-TEX11 Goat Polyclonal Antibody

Supplier: R&D Systems
AF5627
220012-190EA 663.78 USD
220012-190
Anti-TEX11 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TEX11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR