Anti-GFRA1 Goat Polyclonal Antibody

Supplier: R&D Systems
AF560
220012-179EA 721.63 USD
220012-179
Anti-GFRA1 Goat Polyclonal Antibody
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen Symbol: GFRA1
Antigen Name: GDNF family receptor alpha 1
Antigen Synonyms: -
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR