Anti-TGFBR2 Goat Polyclonal Antibody

Supplier: R&D Systems
AF532
220012-049EA 707.17 USD
220012-049
Anti-TGFBR2 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TGFBR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR