Anti-IGF2BP2 Goat Polyclonal Antibody

Supplier: R&D Systems
AF5305
220012-042EA 678.25 USD
220012-042
Anti-IGF2BP2 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IGF2BP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR