Anti-IL10 Goat Polyclonal Antibody

Supplier: R&D Systems
AF519
220012-001EA 736.09 USD
220012-001
Anti-IL10 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR