Anti-SOX18 Sheep Polyclonal Antibody

Supplier: R&D Systems
AF5077
220011-946EA 620.4 USD
220011-946
Anti-SOX18 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SOX18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR