Anti-CALB2 Goat Polyclonal Antibody

Supplier: R&D Systems
AF5065
220011-938EA 736.09 USD
220011-938
Anti-CALB2 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CALB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR