Anti-NEDD8 Sheep Polyclonal Antibody

Supplier: R&D Systems
AF4936
220011-874EA 678.25 USD
220011-874
Anti-NEDD8 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: NEDD8
Antigen Name: neural precursor cell expressed, developmentally down-regulated 8
Antigen Synonyms: NEDD-8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR