Anti-ITM2C Sheep Polyclonal Antibody

Supplier: R&D Systems
AF4904
220011-854EA 678.25 USD
220011-854
Anti-ITM2C Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ITM2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR