Anti-GAB2 Sheep Polyclonal Antibody

Supplier: R&D Systems
AF4877
220011-839EA 649.32 USD
220011-839
Anti-GAB2 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: GAB2
Antigen Name: GRB2-associated binding protein 2
Antigen Synonyms: -
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR