Anti-IL6ST Goat Polyclonal Antibody

Supplier: R&D Systems
AF468
220011-747EA 721.63 USD
220011-747
Anti-IL6ST Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL6ST
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR