Anti-NFKBIE Sheep Polyclonal Antibody

Supplier: R&D Systems
AF4637
220011-725EA 649.32 USD
220011-725
Anti-NFKBIE Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: NFKBIE
Antigen Name: nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, epsilon
Antigen Synonyms: I-kappa-B-epsilon, IKBE, IkB-E, NF-kappa-BIE, ikB-epsilon
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR