Anti-IKBKB Sheep Polyclonal Antibody

Supplier: R&D Systems
AF4535
220011-683EA 692.71 USD
220011-683
Anti-IKBKB Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: IKBKB
Antigen Name: inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta
Antigen Synonyms: IKK-beta, IKK2, IKKB, IMD15, NFKBIKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR