Anti-CCL3 Goat Polyclonal Antibody

Supplier: R&D Systems
AF-450-NA
220011-668EA 750.55 USD
220011-668
Anti-CCL3 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CCL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR