Anti-NT5E Sheep Polyclonal Antibody

Supplier: R&D Systems
AF4488
220011-657EA 750.55 USD
220011-657
Anti-NT5E Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NT5E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR