Anti-PTK2 Sheep Polyclonal Antibody

Supplier: R&D Systems
AF4467
220011-647EA 663.78 USD
220011-647
Anti-PTK2 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PTK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR