Anti-NETO1 Sheep Polyclonal Antibody

Supplier: R&D Systems
AF4337
220011-590EA 678.25 USD
220011-590
Anti-NETO1 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: NETO1
Antigen Name: neuropilin (NRP) and tolloid (TLL)-like 1
Antigen Synonyms: BCTL1, BTCL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR