Anti-FGF8 Goat Polyclonal Antibody

Supplier: R&D Systems
AF-423-NA
220011-541EA 678.25 USD
220011-541
Anti-FGF8 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: FGF8 (b Isoform)
Antigen Name: fibroblast growth factor 8
Antigen Synonyms: Aigf, Fgf-8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR