Anti-FOLH1 Sheep Polyclonal Antibody

Supplier: R&D Systems
AF4234
220011-537EA 707.17 USD
220011-537
Anti-FOLH1 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: FOLH1
Antigen Name: folate hydrolase (prostate-specific membrane antigen) 1
Antigen Synonyms: FGCP, FOLH, GCP2, GCPII, NAALAD1, NAALAdase, PSM, PSMA, mGCP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR