Anti-CYR61 Sheep Polyclonal Antibody

Supplier: R&D Systems
AF4055
220011-442EA 692.71 USD
220011-442
Anti-CYR61 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CYR61
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR