Anti-PPP2R1A Rabbit Polyclonal Antibody

Supplier: R&D Systems
AF3989
220011-404EA 591.48 USD
220011-404
Anti-PPP2R1A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PPP2R1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR