Anti-SERPINE1 Goat Polyclonal Antibody

Supplier: R&D Systems
AF3828
220011-322EA 707.17 USD
220011-322
Anti-SERPINE1 Goat Polyclonal Antibody
Antibodies
Simple Western

Type: Primary
Antigen: SERPINE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR