Anti-ITGA9 Goat Polyclonal Antibody

Supplier: R&D Systems
AF3827
220011-321EA 707.17 USD
220011-321
Anti-ITGA9 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ITGA9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR