Anti-CCL5 Sheep Polyclonal Antibody

Supplier: R&D Systems
AF3819
220011-314EA 649.32 USD
220011-314
Anti-CCL5 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CCL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR