Anti-PRDX5 Goat Polyclonal Antibody

Supplier: R&D Systems
AF3774
220011-288EA 577.02 USD
220011-288
Anti-PRDX5 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PRDX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR