Anti-NRG1 Goat Polyclonal Antibody

Supplier: R&D Systems
AF377
220011-286EA 663.78 USD
220011-286
Anti-NRG1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NRG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Extracellular Domain
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR