Anti-ACVR2A Goat Polyclonal Antibody

Supplier: R&D Systems
AF340
220011-088EA 750.55 USD
220011-088
Anti-ACVR2A Goat Polyclonal Antibody
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: ACVR2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR