Anti-ACVR2B Goat Polyclonal Antibody

Supplier: R&D Systems
AF339
220011-083EA 750.55 USD
220011-083
Anti-ACVR2B Goat Polyclonal Antibody
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: ACVR2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR