Anti-CLEC2D Goat Polyclonal Antibody

Supplier: R&D Systems
AF3376
220011-072EA 678.25 USD
220011-072
Anti-CLEC2D Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CLEC2D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR