Anti-GM12169 Goat Polyclonal Antibody

Supplier: R&D Systems
AF3374
220011-071EA 577.02 USD
220011-071
Anti-GM12169 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: GM12169
Antigen Name: predicted gene 12169
Antigen Synonyms: OTTMUSG00000005497
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR