Anti-TP53 Rabbit Polyclonal Antibody

Supplier: R&D Systems
AF3306
220011-038EA 548.09 USD
220011-038
Anti-TP53 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TP53
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR