Anti-CSF1R Goat Polyclonal Antibody

Supplier: R&D Systems
AF329
220011-029EA 663.78 USD
220011-029
Anti-CSF1R Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CSF1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR