Anti-FAS Goat Polyclonal Antibody

Supplier: R&D Systems
AF326
220011-009EA 721.63 USD
220011-009
Anti-FAS Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: FAS
Antigen Name: Fas cell surface death receptor
Antigen Synonyms: ALPS1A, APO-1, APT1, CD95, FAS1, FASTM, TNFRSF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR