Anti-LILRB5 Goat Polyclonal Antibody

Supplier: R&D Systems
AF3065
220010-903EA 692.71 USD
220010-903
Anti-LILRB5 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LILRB5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR