Anti-SDC4 Goat Polyclonal Antibody

Supplier: R&D Systems
AF2918
220010-815EA 721.63 USD
220010-815
Anti-SDC4 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SDC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR