Anti-FAM3C Goat Polyclonal Antibody

Supplier: R&D Systems
AF2868
220010-791EA 678.25 USD
220010-791
Anti-FAM3C Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: FAM3C
Antigen Name: family with sequence similarity 3, member C
Antigen Synonyms: D6Wsu176e, Ilei
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR