Anti-LUM Goat Polyclonal Antibody

Supplier: R&D Systems
AF2846
220010-774EA 692.71 USD
220010-774
Anti-LUM Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LUM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR