Anti-CD5L Goat Polyclonal Antibody

Supplier: R&D Systems
AF2834
220010-767EA 721.63 USD
220010-767
Anti-CD5L Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD5L
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR