Anti-TIM6 Goat Polyclonal Antibody

Supplier: R&D Systems
AF2737
220010-707EA 577.02 USD
220010-707
Anti-TIM6 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TIM6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR