Anti-BIK Goat Polyclonal Antibody

Supplier: R&D Systems
AF2679
220010-674EA 678.25 USD
220010-674
Anti-BIK Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BIK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR