Anti-CRISP2 Goat Polyclonal Antibody

Supplier: R&D Systems
AF2575
220010-609EA 649.32 USD
220010-609
Anti-CRISP2 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CRISP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR