Anti-CD200R1 Goat Polyclonal Antibody

Supplier: R&D Systems
AF2554
220010-593EA 721.63 USD
220010-593
Anti-CD200R1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD200R1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR