Anti-IL18 Goat Polyclonal Antibody

Supplier: R&D Systems
AF2548
220010-591EA 750.55 USD
220010-591
Anti-IL18 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rhesus Macaque
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR