Anti-PSPN Sheep Polyclonal Antibody

Supplier: R&D Systems
AF2479
220010-544EA 649.32 USD
220010-544
Anti-PSPN Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PSPN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR